• USA Home
  • 46477 - Nandrolone 17-propionate

EMAIL THIS PAGE TO A FRIEND
46477 Sigma-Aldrich

Nandrolone 17-propionate

VETRANAL, analytical standard

  • CAS Number 7207-92-3

  • Empirical Formula (Hill Notation) C21H30O3

  • Molecular Weight 330.46

  •  Beilstein Registry Number 3157236

  •  EC Number 230-587-0

  •  MDL number MFCD00198408

  •  PubChem Substance ID 329757198

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments, Hormones,
grade   analytical standard
InChI Key   LGRKCTFWUWAKON-RRFJAZBJSA-N
product line   VETRANAL
form   neat
shelf life   limited shelf life, expiry date on the label
drug control   USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?