EMAIL THIS PAGE TO A FRIEND
46480 Sigma-Aldrich

3-Fluoroaniline

technical, ≥97.0% (GC)

Synonym: 1-Amino-3-fluorobenzene

Purchase

Properties

Related Categories Aryl Fluorinated Building Blocks, Building Blocks, C6, Chemical Synthesis, Fluorinated Building Blocks,
InChI Key   QZVQQUVWFIZUBQ-UHFFFAOYSA-N
grade   technical
assay   ≥97.0% (GC)
refractive index   n20/D 1.542(lit.)
  n20/D 1.545
bp   186 °C/756 mmHg(lit.)
density   1.156 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2941 6.1 / PGIII
WGK Germany 
3
RTECS 
BY1400000
Flash Point(F) 
170.6 °F
Flash Point(C) 
77 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?