EMAIL THIS PAGE TO A FRIEND
46490 Sigma-Aldrich

4-Fluoroaniline

technical, ≥97.0% (GC)

Synonym: 1-Amino-4-fluorobenzene

Purchase

Properties

Related Categories Aryl Fluorinated Building Blocks, Building Blocks, C6, Chemical Synthesis, Fluorinated Building Blocks,
InChI Key   KRZCOLNOCZKSDF-UHFFFAOYSA-N
grade   technical
assay   ≥97.0% (GC)
refractive index   n20/D 1.539(lit.)
  n20/D 1.540
bp   187 °C/767 mmHg(lit.)
density   1.173 g/mL at 25 °C(lit.)

Description

Application

4-Fluoroaniline may be employed as one of the target pollutant during the investigation of biodegradability of fluorinated compounds under aerobic conditions. It may be used for the synthesis of luminescent and cationic monocyclometalated gold(III) monoaryl complexes.

General description

The barriers to inversion and internal rotation of the NH2 group in 4-fluoroaniline has been investigated by far infrared gas spectra.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2941 6.1 / PGIII
WGK Germany 
3
RTECS 
BY1575000
Flash Point(F) 
165.2 °F
Flash Point(C) 
74 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

F3800 4-Fluoroaniline, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?