• USA Home
  • 46551 - β-Estradiol 17-acetate

EMAIL THIS PAGE TO A FRIEND
46551 Sigma-Aldrich

β-Estradiol 17-acetate

VETRANAL, analytical standard

Synonym: 1,3,5(10)-Estratriene-3,17β-diol 17-acetate, 3,17β-Dihydroxy-1,3,5(10)-estratriene 17-acetate

  • CAS Number 1743-60-8

  • Empirical Formula (Hill Notation) C20H26O3

  • Molecular Weight 314.42

  •  Beilstein Registry Number 2625732

  •  MDL number MFCD00056537

  •  PubChem Substance ID 24870282

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments, Biomarkers and Metabolomics Standards,
grade   analytical standard
InChI Key   QAHOQNJVHDHYRN-SLHNCBLASA-N
product line   VETRANAL
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?