• USA Home
  • 46554 - β-Estradiol 3,17-dipropionate

EMAIL THIS PAGE TO A FRIEND
46554 Sigma-Aldrich

β-Estradiol 3,17-dipropionate

VETRANAL, analytical standard

Synonym: 1,3,5(10)-Estratriene 3,17β-dipropionate, 3,17β-Dihydroxy-1,3,5(10)-estratriene 3,17-dipropionate

  • CAS Number 113-38-2

  • Empirical Formula (Hill Notation) C24H32O4

  • Molecular Weight 384.51

  •  Beilstein Registry Number 3223915

  •  EC Number 204-026-5

  •  MDL number MFCD00051146

  •  PubChem Substance ID 329757211

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments, Biomarkers and Metabolomics Standards,
grade   VETRANAL, analytical standard
InChI Key   JQIYNMYZKRGDFK-RUFWAXPRSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
KG4725000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?