• USA Home
  • 46556 - β-Estradiol 17-propionate

EMAIL THIS PAGE TO A FRIEND
46556 Sigma-Aldrich

β-Estradiol 17-propionate

VETRANAL, analytical standard

Synonym: 1,3,5(10)-Estratriene-3,17β-diol 17-propionate, 3,17β-Dihydroxy-1,3,5(10)-estratriene 17-propionate

  • CAS Number 3758-34-7

  • Empirical Formula (Hill Notation) C21H28O3

  • Molecular Weight 328.45

  •  Beilstein Registry Number 3215697

  •  EC Number 223-164-7

  •  MDL number MFCD00056539

  •  PubChem Substance ID 329757212

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Biomarkers and Metabolomics Standards, Hormone Standards
grade   analytical standard
InChI Key   PQCRZWCSVWBYSC-UHFFFAOYSA-N
product line   VETRANAL
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?