• USA Home
  • 46608 - Procaine hydrochloride

EMAIL THIS PAGE TO A FRIEND
46608 Sigma-Aldrich

Procaine hydrochloride

VETRANAL, analytical standard

Synonym: 4-Aminobenzoic acid 2-diethylaminoethyl ester, p-Aminobenzoic acid diethylaminoethyl ester hydrochloride, Novocaine hydrochloride

  • CAS Number 51-05-8

  • Linear Formula H2NC6H4CO2CH2CH2N(C2H5)2·HCl

  • Molecular Weight 272.77

  •  Beilstein Registry Number 3917802

  •  EC Number 200-077-2

  •  MDL number MFCD00013000

  •  PubChem Substance ID 329757218

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Anesthetics Standards, Antibiotics Standards,
grade   analytical standard
InChI Key   HCBIBCJNVBAKAB-UHFFFAOYSA-N
product line   VETRANAL
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   155-156 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
DG2275000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?