EMAIL THIS PAGE TO A FRIEND
46762 Sigma-Aldrich

Sulfabenzamide

VETRANAL, analytical standard

Synonym: N-(4-Aminobenzenesulfonyl)benzamide, N-Sulfanilylbenzamide

  • CAS Number 127-71-9

  • Empirical Formula (Hill Notation) C13H12N2O3S

  • Molecular Weight 276.31

  •  Beilstein Registry Number 2139003

  •  EC Number 204-859-4

  •  MDL number MFCD00044890

  •  PubChem Substance ID 24870426

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Antibiotics, Antibiotics A to Z,
grade   analytical standard
InChI Key   PBCZLFBEBARBBI-UHFFFAOYSA-N
product line   VETRANAL
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
CV5802500

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?