467901 Sigma-Aldrich

Nickel(II) sulfate hexahydrate

≥99.99% trace metals basis

DOWNLOAD MSDS (PDF)
  • CAS Number 10101-97-0

  • Linear Formula NiSO4 · 6H2O

  • Molecular Weight 262.85

  •  EC Number 232-104-9

  •  MDL number MFCD00149813

  •  PubChem Substance ID 24870445

Properties

vapor density   2.07 (vs air)
assay   ≥99.99% trace metals basis
impurities   ≤0.002% N compounds
  ≤0.005% insolubles
  <100 ppm total metallic impurities
anion traces   chloride (Cl-): ≤0.001%
cation traces   Ca: ≤0.005%
  Co: ≤0.002%
  Cu: ≤0.005%
  Fe: ≤0.001%
  K: ≤0.01%
  Mg: ≤0.005%
  Mn: ≤0.002%
  Na: ≤0.05%

Description

Packaging

50, 250 g in poly bottle

Price and Availability

Customers Also Viewed

Nickel(II) sulfate

anhydrous, 99.99% trace metals basis

Nickel(II) sulfate heptahydrate

99.999% trace metals basis

Nickel(II) sulfate hexahydrate

puriss. p.a., ACS reagent, 99-102.0%

Nickel(II) sulfate hexahydrate

purum p.a., crystallized, ≥99.0% (KT)

Nickel(II) sulfate hexahydrate

ACS reagent, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?