EMAIL THIS PAGE TO A FRIEND
46835 Sigma-Aldrich

2-Fluorobiphenyl

purum, ≥98.0% (GC)

Purchase

Properties

Related Categories Aryl, Aryl Fluorinated Building Blocks, Building Blocks, C8-C12, C9 to C12,
InChI Key   KLECYOQFQXJYBC-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
bp   248 °C(lit.)
mp   71-74 °C(lit.)
  71-74 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

102741 2-Fluorobiphenyl, 96%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?