• USA Home
  • 46978 - Fluorescein Low Range DNA standard

EMAIL THIS PAGE TO A FRIEND
46978 Sigma-Aldrich

Fluorescein Low Range DNA standard

fluorescein-labeled marker for DNA electrophoresis

Synonym: Deoxyribonucleic acid

Purchase

Properties

Related Categories Biochemicals and Reagents, Fluorescent Nucleic Acid and Protein Markers, Fluorescent Probes, Labels, Particles and Stains More...
grade   for molecular biology
InChI Key   GNBHRKFJIUUOQI-UHFFFAOYSA-N
form   liquid
usage   sufficient for 100 tests
suitability   suitable for electrophoresis (DNA)
shipped in   dry ice
storage temp.   −20°C

Description

Components

Fluorescein Low Range DNA Standard is provided as two tubes:
• 1 vial (200 μl volume) pBR 322 Hin F1 digest in 10 mM TRIS (pH 8.0), and 1 mM EDTA (50 fmol/μl each fragment; 0.14 μg/μl)
• 1 vial 5× loading buffer (500 μl volume) consisting of 12.5% Ficoll, 0.05% bromophenol blue, 50 mM TRIS (pH 8.0), and 5 mM EDTA

Application

Suitable for size determination of dsDNA by DNA electrophoresis with either agarose or polyacrylamide gels.

Features and Benefits

• Excellent for UV illumination based imaging
• Excitation at 302 nm
• Can be used with multi-color imaging

General description

Sigma’s Fluorescein Low Range DNA Standard contains 10 fluorescein end-labeled fragments as follows: 75, 154, 220/221, 296, 334, 396, 504, 517, and 1632 bp. The standard should be diluted with the included loading buffer before loading on an agarose or polyacrylamide gel. For detection, the fluorescein should be excited at 302nm for emission at 521 nm.

Other Notes

For optimal resolution, the recommended polyacrylamide gel concentration is < 10% and the optimal agarose gel concentration is > 2.5%, cast < 1mm thick. bp.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?