• USA Home
  • 47110 - 2-Fluoro-1-methylpyridinium p-toluenesulfonate

EMAIL THIS PAGE TO A FRIEND
47110 Sigma-Aldrich

2-Fluoro-1-methylpyridinium p-toluenesulfonate

technical, ≥90% (F)

Purchase

Properties

Related Categories Chemical Synthesis, Coupling, Other Synthetic Reagents for Coupling, Synthetic Reagents
InChI Key   HQWDKLAIDBOLFE-UHFFFAOYSA-M
grade   technical
assay   ≥90% (F)
mp   130-134 °C(lit.)

Description

Other Notes

Efficient coupling reagent. Review; Synthesis of thioesters; Stereoselective synthesis of α-ribonucleotides

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?