• USA Home
  • 47160 - 1-Fluoro-3-nitrobenzene

EMAIL THIS PAGE TO A FRIEND
47160 Sigma-Aldrich

1-Fluoro-3-nitrobenzene

purum, ≥97.0% (GC)

Purchase

Properties

Related Categories Aryl Fluorinated Building Blocks, Building Blocks, C6, Chemical Synthesis, Fluorinated Building Blocks,
InChI Key   WMASLRCNNKMRFP-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
color   green-yellow
refractive index   n20/D 1.525(lit.)
  n20/D 1.525
bp   205 °C(lit.)
mp   1.7 °C(lit.)
density   1.325 g/mL at 25 °C(lit.)

Description

Application

1-Fluoro-3-nitrobenzene may be used in the preparation of N-[6-(3-nitrophenoxy)-1,3-benzothiazol-2-yl]cyclopropanecarboxamide. It may also be used in the synthesis various of aryl–Au(I) compounds.

General description

1-Fluoro-3-nitrobenzene is a m-substituted nitrobenzene derivative. Its toxicity against Tetrahymena pyriformis has been assessed.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
188.6 °F
Flash Point(C) 
87 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?