EMAIL THIS PAGE TO A FRIEND
47428 Sigma-Aldrich

Fmoc-Phe(4-F)-OH

≥98.0% (HPLC)

Synonym: Fmoc-4-fluoro-L-phenylalanine

  • CAS Number 169243-86-1

  • Empirical Formula (Hill Notation) C24H20FNO4

  • Molecular Weight 405.42

  •  Beilstein Registry Number 8287377

  •  MDL number MFCD00191197

  •  PubChem Substance ID 329757314

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Phenylalanine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   IXUMACXMEZBPJG-QFIPXVFZSA-N
assay   ≥98.0% (HPLC)
storage temp.   2-8°C

Description

Packaging

5 g in poly bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?