EMAIL THIS PAGE TO A FRIEND
47434 Sigma-Aldrich

Fmoc-D-Phe(4-NO2)-OH

purum, ≥98.0% (TLC)

Synonym: Fmoc-4-nitro-D-phenylalanine

  • CAS Number 177966-63-1

  • Empirical Formula (Hill Notation) C24H20N2O6

  • Molecular Weight 432.43

  •  Beilstein Registry Number 8873625

  •  MDL number MFCD00237029

  •  PubChem Substance ID 24870990

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Phenylalanine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   RZRRJPNDKJOLHI-JOCHJYFZSA-N
grade   purum
assay   ≥98.0% (TLC)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?