EMAIL THIS PAGE TO A FRIEND
47448 Sigma-Aldrich

Fmoc-Cys(tBu)-OPfp

purum, ≥98.0% (TLC)

Synonym: Nα-Fmoc-S-tert-butyl-L-cysteine pentafluorophenyl ester

  • CAS Number 109434-23-3

  • Empirical Formula (Hill Notation) C28H24F5NO4S

  • Molecular Weight 565.55

  •  Beilstein Registry Number 5842866

  •  MDL number MFCD00065636

  •  PubChem Substance ID 329757317

Purchase

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?