EMAIL THIS PAGE TO A FRIEND
47580 Sigma-Aldrich

5-Fluorouridine

≥99.0% (HPLC)

Synonym: 5-Fluorouracil 1β-D-ribofuranoside, FUrd

  • CAS Number 316-46-1

  • Empirical Formula (Hill Notation) C9H11FN2O6

  • Molecular Weight 262.19

  •  Beilstein Registry Number 33662

  •  EC Number 206-260-3

  •  MDL number MFCD00036832

  •  PubChem Substance ID 329757325

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   FHIDNBAQOFJWCA-UAKXSSHOSA-N
assay   ≥99.0% (HPLC)
optical activity   [α]20/D +18±1°, c = 1% in H2O
mp   182-184 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3
RTECS 
YU8050000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

F5130 5-Fluorouridine, proapoptotic anitproliferative plant growth regulator

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?