• USA Home
  • 47709 - Formate Dehydrogenase from Candida boidinii

EMAIL THIS PAGE TO A FRIEND
47709 Sigma-Aldrich

Formate Dehydrogenase from Candida boidinii

powder, 0.3-0.6 U/mg

Synonym: FDH, Formate:NAD+ oxidoreductase

Purchase

Properties

Related Categories 1.2.x.x Acting on aldehydes or oxo groups, 1.x.x.x Oxidoreductases, Biochemicals and Reagents, Cell Biology, Chemical Synthesis,
form   powder
mol wt   Mr ~76000
foreign activity   alcohol dehydrogenase <0.05%
  lactate dehydrogenase <0.05%
shipped in   wet ice
storage temp.   −20°C

Description

Biochem/physiol Actions

Formate dehydrogenase is involved in the stress response of plants and catalyzes the reduction of NAD+ to NADH.

Other Notes

Characterization; Stereoselective lactone preparation

Unit Definition

1 U corresponds to the amount of enzyme which oxidizes 1 μmol sodium formate (Cat. No. 71539) per minute at pH 7.6 and 25°C

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?