• USA Home
  • 47720 - (Triphenylphosphoranylidene)acetaldehyde

EMAIL THIS PAGE TO A FRIEND
47720 Sigma-Aldrich

(Triphenylphosphoranylidene)acetaldehyde

technical, ≥95% (NT/PNMR)

Synonym: (Formylmethylene)triphenylphosphorane

  • CAS Number 2136-75-6

  • Linear Formula HCOCH=P(C6H5)3

  • Molecular Weight 304.32

  •  Beilstein Registry Number 523797

  •  EC Number 218-375-6

  •  MDL number MFCD00006994

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C-C Bond Formation, C13-C60, Carbonyl Compounds,
InChI Key   CQCAYWAIRTVXIY-UHFFFAOYSA-N
grade   technical
assay   ≥95% (NT/PNMR)
loss   ≤2% loss on drying, 110°C
mp   184-187 °C
  185-188 °C(lit.)

Description

Other Notes

Wittig reagent for the stereoselective synthesis of Z-α,β-unsaturated aldehydes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

280933 (Triphenylphosphoranylidene)acetaldehyde, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?