EMAIL THIS PAGE TO A FRIEND
47900 Sigma-Aldrich

Fumaric acid

puriss., ≥99.5% (T)

  • CAS Number 110-17-8

  • Linear Formula HOOCCH=CHCOOH

  • Molecular Weight 116.07

  •  Beilstein Registry Number 605763

  •  EC Number 203-743-0

  •  MDL number MFCD00002700

Purchase

Properties

Related Categories Acids, Acids & Bases, Allium cepa (Onion), Building Blocks, C1 to C5,
vapor pressure   1.7 mmHg ( 165 °C)
InChI Key   VZCYOOQTPOCHFL-OWOJBTEDSA-N
grade   puriss.
assay   ≥99.5% (T)
autoignition temp.   1364 °F
expl. lim.   40 %
mp   298-300 °C (subl.)(lit.)

Description

Analysis Note

sublimes at 165°C at 1.7 mm pressure (Lit.)

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
1
RTECS 
LS9625000
Flash Point(F) 
446 °F
Flash Point(C) 
230 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Citric Acid Cycle

Acetyl-CoA enters a cycle whose discovery by Hans Krebs 70 years ago marked a milestone in biochemistry. The citric acid cycle is also termed the tricarboxylic acid cycle, TCA cycle or Krebs cycle (s...
BioFiles 2007, 2.6, 16.
Keywords: Addition reactions, Biochemistry, Biofiles, Catalysis, Citric Acid Cycle, Decarboxylations, Dehydration reaction, Hydration reaction, Metabolites, Oxidations, Phosphorylations

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?