EMAIL THIS PAGE TO A FRIEND
47980 Sigma-Aldrich

Fumaronitrile

purum, ≥98.0% (GC)

Synonym: trans-1,2-Dicyanoethylene

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds,
InChI Key   KYPOHTVBFVELTG-OWOJBTEDSA-N
grade   purum
assay   ≥98.0% (GC)
bp   186 °C(lit.)
mp   93-95 °C(lit.)

Description

Other Notes

Precursor of the corresponding amidate, a cross-linking agent for modifying proteins via amidation according to Davies and Stark

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3439 6.1 / PGIII
WGK Germany 
3
RTECS 
LT2300000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?