EMAIL THIS PAGE TO A FRIEND
48040 Sigma-Aldrich

2-Furoic hydrazide

purum, ≥97.0% (NT)

Synonym: 2-Furoic acid hydrazide, 2-Furoylhydrazine

  • CAS Number 3326-71-4

  • Empirical Formula (Hill Notation) C5H6N2O2

  • Molecular Weight 126.11

  •  Beilstein Registry Number 114435

  •  EC Number 222-046-2

  •  MDL number MFCD00003235

Purchase

Properties

InChI Key   SKTSVWWOAIAIKI-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (NT)
mp   77-79 °C(lit.)
  77-80 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
LV1925000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

130443 2-Furoic hydrazide, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?