• USA Home
  • 48200 - 2-Furyl methyl ketone

EMAIL THIS PAGE TO A FRIEND
48200 Sigma-Aldrich

2-Furyl methyl ketone

purum, ≥99.0% (GC)

Synonym: 2-Acetylfuran

  • CAS Number 1192-62-7

  • Empirical Formula (Hill Notation) C6H6O2

  • Molecular Weight 110.11

  •  Beilstein Registry Number 107909

  •  EC Number 214-757-1

  •  MDL number MFCD00003242

  •  PubChem Substance ID 329757359

Purchase

Properties

Related Categories Building Blocks, C4 to C7, Chemical Synthesis, Furans, Heterocyclic Building Blocks More...
InChI Key   IEMMBWWQXVXBEU-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
refractive index   n20/D 1.5070(lit.)
bp   67 °C/10 mmHg(lit.)
mp   26-28 °C(lit.)
  26-28 °C
density   1.098 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
OB3870000
Flash Point(F) 
159.8 °F
Flash Point(C) 
71 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?