• USA Home
  • 48274 - β-Galactosidase from Escherichia coli

EMAIL THIS PAGE TO A FRIEND
48274 Sigma-Aldrich

β-Galactosidase from Escherichia coli

>400 U/mg

Synonym: β-D-Galactoside galactohydrolase, Lactase

Purchase

Description

Unit Definition

1 U corresponds to the amount of enzyme which hydrolyzes 1 μmol 2-nitrophenyl-β-D-galactopyranoside (Cat. No. 73660) per minute at pH 7.8 and 37 °C

Biochem/physiol Actions

β-galactosidase cleaves lactose into its monosaccharide components, glucose and galactose. It also catalyses the transglycosylation of glucose into allolactose, the inducer of β-galactosidase, in a feedback loop.

Physical properties

Tetramer molecular weight 465 kDa (subunits 116.3 kDa each)

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
LW5840000

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

G5635 β-Galactosidase from Escherichia coli, Grade VIII, lyophilized powder, ≥500 units/mg protein

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?