EMAIL THIS PAGE TO A FRIEND
484016 Sigma-Aldrich

Potassium hydroxide

reagent grade, 90%, flakes

Synonym: Caustic potash

Purchase

Properties

Related Categories Acids & Bases, Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Bases,
grade   reagent grade
vapor pressure   1 mmHg ( 719 °C)
assay   90%
form   flakes
mp   361 °C(lit.)
solubility   water: soluble1120 g/L

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Packaging

1 kg in poly bottle

10 kg in fiber drum

Application

Potassium hydroxide (KOH) was used in the following processes:
• Quantification of glutathione (GSH) and glutathione disulfide (GSSG) in biological samples by HPLC.
• Preparation of buffer for use in the Investigation of seed fatty acid (FA) composition, protein, oil and sugar content.
• Stable isotopic profiling study of Caenorhabditis elegans.
Potassium hydroxide may be employed as an etchant for the laser modified glass. In combination with tellurium powder, it mediates the quantitative pinacolization of aromatic carbonyl compounds.

General description

Potassium hydroxide (KOH) is a hygroscopic, water soluble base synthesized by the electrolysis of potassium chloride solution. It is the raw material for the synthesis of many potassium compounds. KOH reacts with CO2 to form potassium carbonate. This property can be utilized to absorb CO2. It participates as catalyst in aldol condensation reactions. KOH anisotropic etching of silicon is used for the fabrication of sub-micron silicon nitride waveguides using conventional contact lithography. Pulverized KOH may be used for the preparation of chalcones.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1813 8 / PGII
WGK Germany 
1
RTECS 
TT2100000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Aldol Condensation Reaction

The aldol condensation is an organic reaction introduced by Charles Wurtz, who first prepared the β-hydroxy aldehyde from acetaldehdye in 1872.1 In an aldol condensation, an enolate ion reacts with a...
Keywords: Addition reactions, Aldol condensation, Aldol reaction, Alkylations, Antibiotics, Asymmetric synthesis, Catalysis, Condensations, Dehydration reaction, Eliminations, Etherifications, Iodinations, Michael Addition, Rearrangements, Suzuki coupling

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?