• USA Home
  • 48630 - Gallic acid monohydrate

EMAIL THIS PAGE TO A FRIEND
48630 Sigma-Aldrich

Gallic acid monohydrate

purum, ACS reagent, ≥98.0% (HPLC)

Synonym: 3,4,5-Trihydroxybenzoic acid monohydrate

  • CAS Number 5995-86-8

  • Linear Formula (HO)3C6H2CO2H · H2O

  • Molecular Weight 188.13

  •  Beilstein Registry Number 2050274

  •  EC Number 205-749-9

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Organic Acids, Synthetic Reagents More...
grade   ACS reagent
  purum
assay   ≥98.0% (HPLC)
ign. residue   ≤0.05%
loss   9-10% loss on drying
mp   252 °C (dec.)(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
LW7525000
Flash Point(F) 
482 °F
Flash Point(C) 
250 °C

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

398225 Gallic acid monohydrate, ACS reagent, ≥98.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?