EMAIL THIS PAGE TO A FRIEND
48808 Sigma-Aldrich

Geranyl bromide

technical, ≥95.0% (T)

Synonym: trans-1-Bromo-3,7-dimethyl-2,6-octadiene

  • CAS Number 6138-90-5

  • Linear Formula (CH3)2C=CHCH2CH2C(CH3)=CHCH2Br

  • Molecular Weight 217.15

  •  Beilstein Registry Number 1703631

  •  EC Number 228-123-7

  •  MDL number MFCD00000243

  •  PubChem Substance ID 24872328

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
InChI Key   JSCUZAYKVZXKQE-JXMROGBWSA-N
grade   technical
assay   ≥95.0% (T)
refractive index   n20/D 1.504(lit.)
  n20/D 1.504
bp   101-102 °C/12 mmHg(lit.)
density   1.094 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

This product has been replaced by 329118-ALDRICH | Geranyl bromide 95%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3334
WGK Germany 
3
Flash Point(F) 
167 °F
Flash Point(C) 
75 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

329118 Geranyl bromide, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?