• USA Home
  • 49098 - β-D-Glucan from barley

EMAIL THIS PAGE TO A FRIEND
49098 Sigma-Aldrich

β-D-Glucan from barley

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates G, Polysaccharide More...
storage temp.   2-8°C

Description

Other Notes

Application in the activity staining of cellulases in polyacrylamide gels

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?