EMAIL THIS PAGE TO A FRIEND
493546 Sigma-Aldrich

Ethyl alcohol, Pure

200 proof, meets USP testing specifications

Synonym: Ethanol, absolute alcohol, non-denatured ethanol

Purchase

Properties

Related Categories Cell Biology, Ethanol (Ethyl Alcohol), Ethanol (Tax-Paid), Multi-Compendial, Nutrition Research,
grade   ACS reagent (Traceable to ACS product listing.)
concentration   200 proof
evapn. residue   ≤0.0025%
bp   78.3 °C
mp   -114 °C
solubility   water: soluble
λ   in H2O
UV absorption   λ: 240 nm Amax: 0.40
  λ: 250 nm Amax: 0.30
  λ: 260 nm Amax: 0.30
  λ: 270 nm Amax: 0.10
  λ: 340 nm Amax: 0.10
suitability   meets USP testing specifications

Description

Frequently Asked Questions

Frequently Asked Questions are available for this Product.

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Sales restrictions may apply

Packaging

1, 4 L in glass bottle

200 L in Pure-Pac™ 2

500, 6×500 mL in glass bottle

Application

Pure ethyl alcohol has been used for the homogenization of tissue samples.

General description

Prices include Federal Tax.

The product is a non-denatured 200proof ethanol. Ethanol, a primary alcohol, is an important raw material for the synthesis of various compounds. It can be synthesized either from molasses by fermentation or by hydration of ethylene. It acts as a hydrogen source and reaction medium during the transfer hydrogenation of carbonyl compounds in the presence of sodium hydroxide. The impact of ethanol/water fumigation on the performance of diesel engines has been studied.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1170 3 / PGII
WGK Germany 
1
RTECS 
KQ6300000
Flash Point(F) 
57.2 °F
Flash Point(C) 
14 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

792799 Ethyl alcohol, Pure, 190 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required

suggested gloves for splash protection

Product #

Description

Add to Cart

Z677353 Dermatril P nitrile gloves, size XS
Z677388 Dermatril P nitrile gloves, size M
Z677396 Dermatril P nitrile gloves, size L
Z677418 Dermatril P nitrile gloves, size XL

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?