494461 Sigma-Aldrich

Tetrahydrofuran

biotech. grade, ≥99.9%, Inhibitor-free

DOWNLOAD MSDS (PDF)
  • CAS Number 109-99-9

  • Empirical Formula (Hill Notation) C4H8O

  • Molecular Weight 72.11

  •  Beilstein Registry Number 102391

  •  EC Number 203-726-8

  •  MDL number MFCD00005356

  •  PubChem Substance ID 24872906

Properties

Related Categories Biotech, Biotech Solvents, Products, Returnable Containers, Solvent by Type,
grade   biotech. grade
vapor density   2.5 (vs air)
vapor pressure   114 mmHg ( 15 °C)
  143 mmHg ( 20 °C)
assay   ≥99.9%
autoignition temp.   610 °F
expl. lim.   11.8 %
impurities   <0.005% peroxides
color   APHA: <10
refractive index   n20/D 1.407(lit.)
bp   65-67 °C(lit.)
mp   −108 °C(lit.)
density   0.889 g/mL at 25 °C(lit.)
λ   H2O reference
UV absorption   λ: 212 nm Amax: 1.0
  λ: 250 nm Amax: 0.180
  λ: 300 nm Amax: 0.020
  λ: 350-400 nm Amax: 0.005

Description

Packaging

10 L in ss drum

2 L in Sure/Seal™

View returnable container options.

Price and Availability

Customers Also Viewed

Tetrahydrofuran

anhydrous, ≥99.9%, inhibitor-free

Tetrahydrofuran

anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%

Tetrahydrofuran

inhibitor-free, CHROMASOLV® Plus, for HPLC, ≥99.9%

Tetrahydrofuran

CHROMASOLV® Plus, for HPLC, ≥99.9%, inhibitor-free

Tetrahydrofuran

spectrophotometric grade, ≥99.5%, inhibitor-free

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?