CAS Number 7446-70-0
Linear Formula AlCl3
Molecular Weight 133.34
EC Number 231-208-1
MDL number MFCD00003422
PubChem Substance ID 24872967
Popular Documents: Specification Sheet (PDF)
| Related Categories | Aluminum, Aluminum Salts, Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals, |
| grade | ReagentPlus® |
| anhydrous | |
| vapor pressure | 1 mmHg ( 100 °C) |
| assay | ≥99.9% trace metals basis |
| mp | 190 °C(lit.) |
1 kg in glass bottle
The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties.1 Aluminum nitride can be synthesized from the elemnts with AlCl3 added to help facilitate crystallization.2
ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?