• USA Home
  • 49627 - Glutaraldehyde solution

EMAIL THIS PAGE TO A FRIEND
49627 Sigma-Aldrich

Glutaraldehyde solution

for electron microscopy, ~8% in H2O

Synonym: Glutaric dialdehyde solution, Pentane-1,5-dial

  • CAS Number 111-30-8

  • Linear Formula OHC(CH2)3CHO

  • Molecular Weight 100.12

  •  Beilstein Registry Number 605390

  •  MDL number MFCD00007025

  •  PubChem Substance ID 329757493

Purchase

Properties

grade   for electron microscopy
InChI Key   SXRSQZLOMIGNAQ-UHFFFAOYSA-N
concentration   ~8% in H2O
impurities   ≤0.05% free carboxylic acid
refractive index   n20/D 1.345
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

G7526 Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?