• USA Home
  • 49632 - Glutaraldehyde solution

EMAIL THIS PAGE TO A FRIEND
49632 Sigma-Aldrich

Glutaraldehyde solution

for electron microscopy, ~25% in H2O

Synonym: Glutaric dialdehyde solution, Pentane-1,5-dial

  • CAS Number 111-30-8

  • Linear Formula OHC(CH2)3CHO

  • Molecular Weight 100.12

  •  Beilstein Registry Number 605390

  •  MDL number MFCD00007025

  •  PubChem Substance ID 329757497

Purchase

Properties

grade   for electron microscopy
InChI Key   SXRSQZLOMIGNAQ-UHFFFAOYSA-N
packaging   pkg of 10 × 2 mL
concentration   ~25% in H2O
impurities   ≤0.05% free carboxylic acid
refractive index   n20/D 1.373
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Supplemental Hazard Statements 
Corrosive to the respiratory tract.
RIDADR 
UN3265 - class 8 - PG 2 - EHS - Corrosive liquid, acidic, organic, n.o.s., HI: all
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

G5882 Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?