EMAIL THIS PAGE TO A FRIEND
49710 Sigma-Aldrich

Glutaryl chloride

technical, ≥97.0% (T)

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
InChI Key   YVOFTMXWTWHRBH-UHFFFAOYSA-N
grade   technical
assay   ≥97.0% (T)
refractive index   n20/D 1.472(lit.)
  n20/D 1.472
bp   216-218 °C(lit.)
density   1.315 g/mL at 20 °C
  1.324 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2922 6.1(8) / PGIII
WGK Germany 
3
RTECS 
SA2583000
Flash Point(F) 
224.6 °F
Flash Point(C) 
107 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

G4608 Glutaryl chloride, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?