• USA Home
  • 50036 - DL-Glyceric acid hemicalcium salt hydrate

EMAIL THIS PAGE TO A FRIEND
50036 Sigma-Aldrich

DL-Glyceric acid hemicalcium salt hydrate

puriss., ≥99.0% (NT), 1-2 mol/mol water

Synonym: 2,3-Dihydroxypropanoic acid hemicalcium salt, Hemicalcium DL-glycerate, hemi-Calcium DL-glycerate

  • CAS Number 207300-72-9

  • Linear Formula [HOCH2CH(OH)COO]2Ca · ×H2O

  • Molecular Weight 250.22 (anhydrous basis)

  •  Beilstein Registry Number 4614104

  •  EC Number 282-247-6

  •  MDL number MFCD00149289

Purchase

Properties

Related Categories Carbohydrate Chemistry, Carbohydrate Derivatives, Chemical Biology, Chemical Synthesis
InChI Key   OKEVQKKVYQHNFW-UHFFFAOYSA-L
grade   puriss.
assay   ≥99.0% (NT)
impurities   1-2 mol/mol water
mp   134 °C (dec.)(lit.)

Description

Other Notes

Preparation of the free acid from the salt

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

G7274 DL-Glyceric acid hemicalcium salt hydrate, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?