• USA Home
  • 50039 - Glycidyl isopropyl ether

EMAIL THIS PAGE TO A FRIEND
50039 Sigma-Aldrich

Glycidyl isopropyl ether

purum, ≥98.0% (GC)

Synonym: 2,3-Epoxypropyl isopropyl ether

  • CAS Number 4016-14-2

  • Empirical Formula (Hill Notation) C6H12O2

  • Molecular Weight 116.16

  •  Beilstein Registry Number 103028

  •  EC Number 223-672-9

  •  MDL number MFCD00005139

  •  PubChem Substance ID 24873038

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Epoxides, Organic Building Blocks, Oxygen Compounds More...
InChI Key   NWLUZGJDEZBBRH-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.410(lit.)
  n20/D 1.410
bp   131-132 °C(lit.)
density   0.924 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
May form explosive peroxides.
RIDADR 
UN 3271 3 / PGIII
WGK Germany 
2
RTECS 
TZ3500000
Flash Point(F) 
95 °F
Flash Point(C) 
35 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?