EMAIL THIS PAGE TO A FRIEND
50128 Sigma-Aldrich

Diethyl oxalate

analytical standard

  • CAS Number 95-92-1

  • Linear Formula C2H5OCOCOOC2H5

  • Molecular Weight 146.14

  •  Beilstein Registry Number 606350

  •  EC Number 202-464-1

  •  MDL number MFCD00009119

  •  PubChem Substance ID 329757570

Purchase

Properties

Related Categories A-L, Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
vapor density   5.03 (vs air)
vapor pressure   1 mmHg ( 47 °C)
InChI Key   WYACBZDAHNBPPB-UHFFFAOYSA-N
assay   ≥99.7% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.410(lit.)
  n20/D 1.410
bp   185 °C(lit.)
mp   −41 °C(lit.)
density   1.076 g/mL at 25 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Kidney
RIDADR 
UN 2525 6.1 / PGIII
WGK Germany 
1
RTECS 
RO2800000
Flash Point(F) 
167 °F
Flash Point(C) 
75 °C
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?