EMAIL THIS PAGE TO A FRIEND
50370 Sigma-Aldrich

Gly-Phe

puriss., ≥99.0% (NT)

Synonym: Glycyl-L-phenylalanine

  • CAS Number 3321-03-7

  • Linear Formula NH2CH2CONHCH(COOH)CH2C6H5

  • Molecular Weight 222.24

  •  Beilstein Registry Number 2279318

  •  EC Number 222-027-9

  •  MDL number MFCD00065110

  •  PubChem Substance ID 24873359

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Poly Amino Acids More...
InChI Key   JBCLFWXMTIKCCB-VIFPVBQESA-N
grade   puriss.
assay   ≥99.0% (NT)
optical activity   [α]20/D +42±2°, c = 2% in H2O
mp   ~264 °C (dec.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?