EMAIL THIS PAGE TO A FRIEND
50524 Sigma-Aldrich

Agar

anti-swarming

Synonym: Agar-agar, Gum agar

Purchase

Properties

Related Categories Agars, Base Ingredients for Media, Microbiology More...
form   powder
shelf life   limited shelf life, expiry date on the label
ign. residue   ≤3.2%
loss   ≤10% loss on drying
turbidity   ≤8 NTU (after autocalving)
pH   6.0-7.5 (1.5% in gel, after autoclaving)
mp   85-95 °C
transition temp   gel point 33-36 °C
gel strength   ≤1050 g/cm2
cation traces   Ca: ≤3000 mg/kg
  Mg: ≤900 mg/kg

Description

Analysis Note

toxic substances: absence
swarming test: positive

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
AW7950000

Documents

Certificate of Analysis

Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?