• USA Home
  • 50586 - Glycol methacrylate kit

EMAIL THIS PAGE TO A FRIEND
50586 Sigma-Aldrich

Glycol methacrylate kit

Synonym: 2-Hydroxyethyl methacrylate kit

Purchase

Description

Components

Composition:
64168 2-Hydroxyethyl methacrylate, 2×250 ml
32250 Diethylene glycol monobutyl ether, 100 ml
33581 Benzoyl peroxide, 10 g
81170 Polyethylene glycol 400, 2×5 ml
39440 N,N-Dimethylaniline, 1 ml

Application

Suitable for most staining methods, silver-gold methods, histochemistry and immuno histochemistry. Embedding medium based on glycol methacrylate.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?