EMAIL THIS PAGE TO A FRIEND
50660 Sigma-Aldrich

Glyoxal solution

~40% in H2O (~8.8 M)

Synonym: Ethanedial, Oxalaldehyde

  • CAS Number 107-22-2

  • Linear Formula OHCCHO

  • Molecular Weight 58.04

  •  Beilstein Registry Number 1732463

  •  MDL number MFCD00006957

  •  PubChem Substance ID 329757620

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C1 to C6, Carbonyl Compounds, Chemical Synthesis,
InChI Key   LEQAOMBKQFMDFZ-UHFFFAOYSA-N
concentration   ~40% in H2O (~8.8 M)
refractive index   n20/D 1.409

Description

Caution

May precipitate on storage; redissolve at 50-60 °C.

may precipitate on storage; redissolve at 50-60°C

Other Notes

This form of glyoxal is composed of 3 moles of glyoxal and 2 moles of water in a relatively stable configuration.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?