• USA Home
  • 50710 - Glyoxylic acid monohydrate

EMAIL THIS PAGE TO A FRIEND
50710 Sigma-Aldrich

Glyoxylic acid monohydrate

purum, ≥97.0% (T)

Synonym: Formylformic acid, Oxoethanoic acid

  • CAS Number 563-96-2

  • Linear Formula HCOCO2H · H2O

  • Molecular Weight 92.05

  •  Beilstein Registry Number 969535

  •  EC Number 206-058-5

  •  MDL number MFCD00149497

  •  PubChem Substance ID 329757621

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C1 to C5, C1 to C6, Carbonyl Compounds,
InChI Key   MOOYVEVEDVVKGD-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (T)
bp   100 °C(lit.)
mp   49-52 °C(lit.)
  ~57 °C
solubility   H2O: 0.1 g/mL clear
storage temp.   2-8°C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
1
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?