• USA Home
  • 50932 - Guanidine phosphate monobasic

EMAIL THIS PAGE TO A FRIEND
50932 Sigma-Aldrich

Guanidine phosphate monobasic

≥98.0% (T)

  • CAS Number 5423-22-3

  • Linear Formula NH2C(=NH)NH2 · H3PO4

  • Molecular Weight 157.07

  •  Beilstein Registry Number 3709508

  •  EC Number 226-551-9

  •  MDL number MFCD00066148

  •  PubChem Substance ID 329757652

Purchase

Properties

InChI Key   CEDDGDWODCGBFQ-UHFFFAOYSA-N
assay   ≥98.0% (T)
mp   128-131 °C
solubility   H2O: soluble0.1 g/mL, clear, colorless

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?