• USA Home
  • 51022 - 3-Guanidinopropionic acid

EMAIL THIS PAGE TO A FRIEND
51022 Sigma-Aldrich

3-Guanidinopropionic acid

puriss., ≥99.0% (NT)

Synonym: β-Guanidinopropionic acid

  • CAS Number 353-09-3

  • Linear Formula HN=C(NH2)NHCH2CH2COOH

  • Molecular Weight 131.13

  •  Beilstein Registry Number 1705262

  •  EC Number 206-530-0

  •  MDL number MFCD00045939

Purchase

Properties

Related Categories Arginine Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   KMXXSJLYVJEBHI-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (NT)
impurities   ≤0.5% water
ign. residue   ≤0.05%
mp   222 °C (dec.)(lit.)

Description

Other Notes

Analogue of creatine; Analogue of GABA

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
AY3157500

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

G6878 3-Guanidinopropionic acid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?