• USA Home
  • 51055 - Guanosine 3′,5′-cyclic monophosphate sodium salt

EMAIL THIS PAGE TO A FRIEND
51055 Sigma-Aldrich

Guanosine 3′,5′-cyclic monophosphate sodium salt

≥99.0% (HPLC)

Synonym: 3′,5′-Cyclic GMP monosodium salt, 3′,5′-cGMP-Na, Guanosine 3′,5′-cyclophosphate monosodium salt, cGMP, cyclic GMP

  • CAS Number 40732-48-7

  • Empirical Formula (Hill Notation) C10H11N5NaO7P

  • Molecular Weight 367.19

  •  Beilstein Registry Number 6467425

  •  EC Number 255-056-0

  •  MDL number MFCD00038424

  •  PubChem Substance ID 329757663

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleosides, Nucleotides, Oligonucleotides, Nucleotides More...
InChI Key   KMPIYXNEROUNOG-GWTDSMLYSA-M
assay   ≥99.0% (HPLC)
shipped in   wet ice
storage temp.   −20°C

Description

Biochem/physiol Actions

An important second messenger, cGMP is a major intracellular mediator of extracellular signals such as nitric oxide and natriuretic peptides. Cyclic GMP interacts with three types of intracellular receptor proteins: cGMP-dependent protein kinases, cGMP-regulated channels, and cGMP-regulated cyclic nucleotide phosphodiesterases. A primary action of elevated cGMP levels in vivo is the stimulation of cGMP-dependent protein kinase (PKG).

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

G6129 Guanosine 3′,5′-cyclic monophosphate sodium salt, ≥99% (HPLC), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?