EMAIL THIS PAGE TO A FRIEND
51260 Sigma-Aldrich

Hematoxylin

for microscopy (Hist.), indicator (pH 5.0-6.0)

Synonym: Natural Black 1

  • CAS Number 517-28-2

  • Empirical Formula (Hill Notation) C16H14O6 · xH2O

  • Molecular Weight 302.28 (anhydrous basis)

  •  Beilstein Registry Number 91400

  •  Colour Index Number 75290

  •  EC Number 208-237-3

  •  MDL number MFCD00078111

  •  PubChem Substance ID 329757676

Purchase

Properties

Related Categories Cell Biology, G-H-I, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
grade   for microscopy (Hist.)
  indicator (pH 5.0-6.0)
InChI Key   WZUVPPKBWHMQCE-XJKSGUPXSA-N
mp   200 °C (dec.)(lit.)
Gene Information   rat ... Adora1(29290), Adora2a(25369)

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
MH7875000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?