EMAIL THIS PAGE TO A FRIEND
51302 Sigma-Aldrich

Hafnium(IV) chloride

purum, ≥97% (AT)

Purchase

Properties

vapor pressure   1 mmHg ( 190 °C)
InChI Key   PDPJQWYGJJBYLF-UHFFFAOYSA-J
grade   purum
assay   ≥97% (AT)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 3260 8/PG 2
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?