EMAIL THIS PAGE TO A FRIEND
51531 Sigma-Aldrich

Hentetracontane

analytical standard

  • CAS Number 7194-87-8

  • Linear Formula CH3(CH2)39CH3

  • Molecular Weight 577.11

  •  Beilstein Registry Number 4256786

  •  EC Number 230-561-9

  •  MDL number MFCD00015551

  •  PubChem Substance ID 24873812

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Chemical Class, Fuels and Hydrocarbons,
grade   analytical standard
InChI Key   IMQDQLFUWRBGGN-UHFFFAOYSA-N
assay   ≥98.0% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   83-85 °C
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability


Nylon Membrane and Net Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?