• USA Home
  • 51541 - Heparan sulfate sodium salt from bovine kidney

EMAIL THIS PAGE TO A FRIEND
51541 Sigma-Aldrich

Heparan sulfate sodium salt from bovine kidney

Synonym: Heparitin sulfate sodium salt

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates H-L, Polysaccharide More...
impurities   ~3% total nitrogen (N)
solubility   H2O: soluble1 mg/mL, clear, colorless
storage temp.   2-8°C

Description

Biochem/physiol Actions

Constituent of membrane-associated proteoglycans suggested to contribute to cell-cell adhesion

Other Notes

Antithrombotic activity in vivo; Structure and function of heparan sulphate proteoglycans, review

Sales restrictions may apply

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?