EMAIL THIS PAGE TO A FRIEND
51580 Sigma-Aldrich

Heptadecane

purum, ≥98.0% (GC)

  • CAS Number 629-78-7

  • Linear Formula CH3(CH2)15CH3

  • Molecular Weight 240.47

  •  Beilstein Registry Number 1738898

  •  EC Number 211-108-4

  •  MDL number MFCD00009002

  •  PubChem Substance ID 329757715

Purchase

Properties

Related Categories Acyclic, Alkanes, Aloe Vera, Building Blocks, Cell Biology,
vapor density   8.3 (vs air)
vapor pressure   1 mmHg ( 115 °C)
InChI Key   NDJKXXJCMXVBJW-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.436(lit.)
bp   302 °C(lit.)
mp   20-22 °C(lit.)
density   0.777 g/mL at 25 °C(lit.)

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
MI3550000
Flash Point(F) 
300.2 °F
Flash Point(C) 
149 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

128503 Heptadecane, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?